MilliporeSigma
  • Vasodilatation effect of farnesylacetones, active constituents of Sargassum siliquastrum, on the basilar and carotid arteries of rabbits.
Vasodilatation effect of farnesylacetones, active constituents of Sargassum siliquastrum, on the basilar and carotid arteries of rabbits.
Bioorganic & medicinal chemistry letters (2008-11-14)
Byong-Gon Park, Seong-Chun Kwon, Gab-Man Park, Jungyeob Ham, Woon-Seob Shin, Seokjoon Lee
ABSTRACT
Two farnesylacetones, 311 and 312, major active constituents of Sargassum siliquastrum collected from the coast of the East Sea in Korea, showed a moderate vasodilatation effect on the basilar arteries of rabbits. Therefore, treatment with farnesylacetones 311 and 312 may selectively accelerate cerebral blood flow through dilatation of the basilar artery.