MilliporeSigma
Determination of phenoxyacid herbicides in water.
Journal of chromatography (1991-06-28)
R B Geerdink, A M Graumans, J Viveen
ABSTRACT
Novel clean-up techniques for a polymeric precolumn (PLRP-S) for the subsequent determination of bentazone and eight phenoxy acid herbicides in surface water samples are described. After preconcentration of the components at pH 3 on a 10 x 2 mm I.D. precolumn, the technique consists of a clean-up with 1000 microliters of 0.1 mol/l sodium hydroxide solution (pH 12.5) and of a heartcut consisting of four precolumn bed volumes of eluent directed to waste followed by ten precolumn bed volumes of eluent directed to the analytical column. Analytical separation is performed with acetonitrile-water (30:70) containing 0.005 mol/l of tetrabutylammonium hydrogensulphate (pH 8.3) (which is also the desorption eluent during heartcutting) on a polymeric analytical column (PLRP-S). With 25 ml of surface water, spiked at 0.25 and 1 microgram/l, applied to the precolumn, recoveries for all components were over 85% with a relative standard deviation (n = 5) of ca. 9% at 0.25 microgram/l and ca. 2% at 1 microgram/l. Detection limits in surface water samples are 0.05-0.1 microgram/l. Owing to automation, the total analysis time is ca. 30 min.
Product Description
Supelco
2-(2,4,5-Trichlorophenoxy)propionic acid, PESTANAL®, analytical standard