MilliporeSigma
  • An expeditious and environmentally friendly synthesis of 3-substituted isocoumarins using microwave irradiation.
An expeditious and environmentally friendly synthesis of 3-substituted isocoumarins using microwave irradiation.
Natural product research (2008-10-16)
Khalid Mohammed Khan, Sumbul Ahmed, Zulfiqar Ali Khan, Mubeen Rani, Shahnaz Perveen, Wolfgang Voelter
ABSTRACT
A microwave-assisted, environmentally friendly, high-yielding, time-saving synthesis of medicinally important 3-substituted isocoumarins was carried out in a single step by direct condensation of homophthalic acid with aryol and acyl chlorides under solvent-free conditions without any solid support. The synthesised isocoumarins were structurally characterised by microanalysis, 1H NMR, EI, IR and UV.
Sigma-Aldrich
Homophthalic acid, 98%