MilliporeSigma
  • Definitive risk factors for anastomotic leaks in elective open colorectal resection.
Definitive risk factors for anastomotic leaks in elective open colorectal resection.
Archives of surgery (Chicago, Ill. : 1960) (2008-09-17)
Paul Suding, Erin Jensen, Murray A Abramson, Kamal Itani, Samuel Eric Wilson
ABSTRACT
Anastomotic leaks following elective colorectal resections increase morbidity, mortality, and the need for additional interventions. An accurate understanding of risk factors would potentially reduce anastomotic leaks and/or allow appropriate selection of patients for diverting stomas. Prospective review of patient and operative characteristics that contribute to anastomotic leaks. Fifty-one sites within the United States (May 2002-March 2005). Six hundred seventy-two patients who participated in a trial comparing preoperative antimicrobials in elective open colorectal surgery. Anastomotic leaks were diagnosed using clinical findings and were confirmed with imaging. We examined 20 variables possibly affecting anastomotic healing in univariate and multivariate analyses. There were 24 anastomotic leaks in 672 patients (3.6%) undergoing elective colorectal resection. There were 10 deaths (1.5%). A baseline albumin level of less than 3.5 g/dL (to convert to grams per liter, multiply by 10) (P = .04) and male sex (P = .03) were associated with anastomotic leaks in both univariate and multivariate analyses (adjusted odds ratios, 2.56 and 3.12, respectively). Increased duration of surgery (SD, 60 minutes; odds ratio, 1.53; 95% confidence interval, 1.06-2.22; P = .03) and steroid use at the time of surgery (odds ratio, 3.85; 95% confidence interval, 1.24-11.93; P = .02) were significant in univariate analysis. Surgical procedure with rectal resection; prophylaxis with ertapenem (vs cefotetan); or history of obesity, tobacco use, or diabetes was not associated with anastomotic leaks. Significant risk factors for anastomotic leaks include low preoperative serum albumin level, steroid use, male sex, and increased duration of surgery. Appreciation of risk factors provides a rational basis for temporary diversion.