MilliporeSigma
  • Design, synthesis and characterization of podocarpate derivatives as openers of BK channels.
Design, synthesis and characterization of podocarpate derivatives as openers of BK channels.
Bioorganic & medicinal chemistry letters (2008-09-16)
Yong-Mei Cui, Eriko Yasutomi, Yuko Otani, Takashi Yoshinaga, Katsutoshi Ido, Kohei Sawada, Tomohiko Ohwada
ABSTRACT
We found that the podocarpic acid structure provides a new scaffold for chemical modulators of large-conductance calcium-activated K(+) channels (BK channels). Structure-activity analysis indicates the importance of both the arrangement (i.e., location and orientation) of the carboxylic acid functionality of ring A and the hydrophobic region of ring C for expression of BK channel-opening activity.
Sigma-Aldrich
Podocarpic acid, 98%