MilliporeSigma
  • Photochemical cis-trans isomerization of furylacryloylpeptides and their different kinetic behavior as substrates for carboxypeptidase Y.
Photochemical cis-trans isomerization of furylacryloylpeptides and their different kinetic behavior as substrates for carboxypeptidase Y.
Analytical biochemistry (1991-04-01)
A Kanstrup, O Buchardt
ABSTRACT
Furylacryloyl substrates used in kinetic measurements of proteolytic enzymes are shown to cis-trans photoisomerize quickly in plain daylight. The enzymatic transformation of the two forms is sufficiently different to cause problems for such measurements without careful protection against light.