MilliporeSigma
  • Dissipation behavior of lufenuron, benzoylphenylurea insecticide, in/on Chinese cabbage applied by foliar spraying under greenhouse conditions.
Dissipation behavior of lufenuron, benzoylphenylurea insecticide, in/on Chinese cabbage applied by foliar spraying under greenhouse conditions.
Bulletin of environmental contamination and toxicology (2008-07-02)
S Khay, J H Choi, A M Abd El-Aty, M I R Mamun, B J Park, A Goudah, H C Shin, J H Shim
ABSTRACT
Chinese cabbage has long been consumed as a staple food by the Koreans in various forms of fresh, salted, or fermented as kimchi. To fulfill the off-season demand for this crop, it has become a common practice to be cultivated under greenhouse conditions. Since pesticide residues in/on leafy vegetables have strongly concerned with food safety in the Korean society, the changes of lufenuron residues, in/on Chinese cabbage applied by foliar spraying under greenhouse conditions was investigated. Lufenuron 5% emulsifiable concentrate (EC) was sprayed with diluted solution of recommended and double doses to the crop. The shoots of the cabbage were harvested immediately after spraying, and sequentially the harvests were conveyed to analyze the residual amounts. The deposited level of the analyte in/on Chinese cabbage under greenhouse conditions seemed to be difficult to produce the crop with 0.2 ppm of maximum residue limit (MRL) of the Korea Food and Drug Administration (KFDA).
Product Description
Supelco
Lufenuron, PESTANAL®, analytical standard
Lufenuron for peak identification, European Pharmacopoeia (EP) Reference Standard
Lufenuron, European Pharmacopoeia (EP) Reference Standard