MilliporeSigma
  • [The effect of diabetogenic substances on the zinc and calcium content in the pancreatic islet cells of rabbits].
[The effect of diabetogenic substances on the zinc and calcium content in the pancreatic islet cells of rabbits].
Biulleten' eksperimental'noi biologii i meditsiny (1991-02-01)
E D Gol'dberg, V A Eshchenko, V D Bovt
ABSTRACT
The injections of diabetogenic drugs to rabbits induced a decrease in zinc and calcium content in B insulocytes dependent on the degree of these cells damage and hyperglycemia. The content of these metals in A insulocytes was not significantly altered due to a light injury of these cells. A high concentration of calcium in islet A cells is suggested to be a protective factor to the action of alteration agents.