MilliporeSigma
  • Effects of 2-hydroxy-4-methylthiobutyrate on portal plasma flow and net portal appearance of amino acids in piglets.
Effects of 2-hydroxy-4-methylthiobutyrate on portal plasma flow and net portal appearance of amino acids in piglets.
Amino acids (2008-06-06)
Z F Fang, J Luo, Z L Qi, F R Huang, S J Zhao, M Y Liu, S W Jiang, J Peng
ABSTRACT
To determine whether portal plasma flow (PPF) and net portal appearance of amino acids (AA) could be affected by 2-hydroxy-4-methylthiobutyrate (HMB), six barrows (35-day-old, 8.6+/-1.4 kg), implanted with arterial, portal and mesenteric catheters, were fed a DL-methionine (as the control) or HMB-supplemented diet once hourly and infused intramesenterically with 1% p-amino hippurate. PPF was numerically 9% higher (P=0.09) in HMB-fed pigs than in controls over a 4-6 h period. Compared with controls, pigs fed the HMB diet had increased (P<0.05) net portal balance and/or appearance of leucine, isoleucine, histidine, arginine and alanine, but had decreased (P<0.05) portal appearance of glutamate over a 6-h period. The concentration of acetate in the lumen of the distal small intestine was higher (P=0.01) in HMB-fed pigs than in controls (25.14 vs. 7.64 mmol/kg). mRNA levels for proglucagon and endothelial nitric-oxide synthase (eNOS) in stomach and proximal small intestine, and mRNA levels for GLP-2 receptor (GLP-2R) in stomach were higher (P<0.05) in HMB-fed pigs compared with those in controls. Collectively, HMB supplementation increased concentrations of short-chain fatty acids in intestinal lumen, expression of proglucagon, GLP-2R, and eNOS genes, and net portal absorption of AA. These novel findings from the study with pigs may also have important implications for intestinal nutrition and health in humans.
Sigma-Aldrich
2-Hydroxy-4-(methylthio)butyric acid calcium salt, ≥99.0% (NT)