MilliporeSigma
  • Trafficking properties and activity regulation of the neuronal glycine transporter GLYT2 by protein kinase C.
Trafficking properties and activity regulation of the neuronal glycine transporter GLYT2 by protein kinase C.
The Biochemical journal (2008-03-18)
Amparo Fornés, Enrique Núñez, Pablo Alonso-Torres, Carmen Aragón, Beatriz López-Corcuera
ABSTRACT
The neuronal glycine transporter GLYT2 controls the availability of the neurotransmitter in glycinergic synapses, and the modulation of its function may influence synaptic transmission. The active transporter is located in membrane rafts and reaches the cell surface through intracellular trafficking. In the present study we prove that GLYT2 constitutively recycles between the cell interior and the plasma membrane by means of a monensin-sensitive trafficking pathway. Also, a regulated trafficking can be triggered by PMA. We demonstrate that PMA inhibits GLYT2 transport by causing net accumulation of the protein in internal compartments through an increase of the internalization rate. In addition, a small increase of plasma membrane delivery and a redistribution of the transporter to non-raft domains is triggered by PMA. A previously identified phorbol-ester-resistant mutant (K422E) displaying an acidic substitution in a regulatory site, exhibits constitutive traffic but, in contrast with the wild-type, fails to show glycine uptake inhibition, membrane raft redistribution and trafficking modulation by PMA. We prove that the action of PMA on GLYT2 involves PKC (protein kinase C)-dependent and -independent pathways, although an important fraction of the effects are PKC-mediated. We show the additional participation of signalling pathways triggered by the small GTPase Rac1 on PMA action. GLYT2 inhibition by PMA and monensin also take place in brainstem primary neurons and synaptosomes, pointing to a GLYT2 trafficking regulation in the central nervous system.
Product Description
Supelco
PMA hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®