MilliporeSigma
  • Combination of bevacizumab and docetaxel in docetaxel-pretreated hormone-refractory prostate cancer: a phase 2 study.
Combination of bevacizumab and docetaxel in docetaxel-pretreated hormone-refractory prostate cancer: a phase 2 study.
European urology (2008-02-16)
Giuseppe Di Lorenzo, William D Figg, Sophie D Fossa, Vincenzo Mirone, Riccardo Autorino, Nicola Longo, Ciro Imbimbo, Sisto Perdonà, Antonio Giordano, Mario Giuliano, Roberto Labianca, Sabino De Placido
ABSTRACT
Although the taxanes represent the most active agents for the first-line treatment of metastatic hormone-refractory prostate cancer (HRPC), most patients eventually progress while receiving taxane-based treatments. No agents are approved for second-line therapy in HRPC, but common standard practice for the oncologists is to treat patients also after docetaxel failure. Twenty highly pretreated patients with HRPC received bevacizumab (10mg/kg) and docetaxel (60mg/m(2)) every 3 wk. All patients had bone metastases and eight had measurable lesions. Eleven patients (55%) had major prostate-specific antigen (PSA) responses, and 3 (37.5%) had objective responses. Seven major PSA responses were recorded in the same patients who had reported a >50% PSA decrease after first-line docetaxel. However, four major PSA responses were observed in patients previously nonresponsive to docetaxel alone. The treatment was well tolerated. Our results show that the combination of bevacizumab and docetaxel is active and well tolerated. Continued investigation of bevacizumab with cytotoxic chemotherapy is warranted in HRPC.
Product Description
Docetaxel trihydrate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Pricing and availability is not currently available.