MilliporeSigma
  • Agonist-specific regulation of mu-opioid receptor desensitization and recovery from desensitization.
Agonist-specific regulation of mu-opioid receptor desensitization and recovery from desensitization.
Molecular pharmacology (2008-01-17)
Michael S Virk, John T Williams
ABSTRACT
Agonist-selective actions of opioids on the desensitization of mu-opioid receptors (MORs) have been well characterized, but few if any studies have examined agonist-dependent recovery from desensitization. The outward potassium current induced by several opioids was studied using whole-cell voltage-clamp recordings in locus ceruleus neurons. A brief application of the irreversible opioid antagonist beta-chlornaltrexamine (beta-CNA) was applied immediately after treatment of slices with saturating concentrations of opioid agonists. This approach permitted the measurement of desensitization and recovery from desensitization using multiple opioid agonists, including [Met](5)enkephalin (ME), [d-Ala(2),N-Me-Phe(4),Gly(5)-ol]-enkephalin (DAMGO), etorphine, fentanyl, methadone, morphine, morphine-6-glucuronide, oxycodone, and oxymorphone. The results indicate that desensitization protects receptors from irreversible antagonism with beta-CNA. The amount of desensitization was measured as the decrease in current during a 10-min application of a saturating agonist concentration and was a good predictor of the extent of receptor protection from irreversible inactivation with beta-CNA. After desensitization with ME or DAMGO and treatment with beta-CNA, there was an initial profound inhibition of MOR-induced current that recovered significantly after 45 min. There was, however, no recovery of MOR-mediated current with time after treatment with agonists that did not cause desensitization, such as oxycodone. These results demonstrate that desensitization prevents irreversible inactivation of receptors by beta-CNA.