MilliporeSigma
  • Lack of effect of paracetamol on the pharmacokinetics and metabolism of codeine in man.
Lack of effect of paracetamol on the pharmacokinetics and metabolism of codeine in man.
European journal of clinical pharmacology (1991-01-01)
A Somogyi, F Bochner, Z R Chen
ABSTRACT
Plasma and urine concentrations of codeine and its measurable metabolites were determined by HPLC in six healthy subjects after a single 30 mg oral dose of codeine either alone or after 7 doses of 1 g paracetamol 8 hourly. After codeine alone, the t1/2 (h), AUC (mumol.l-1.h) and CLR (ml.min-1) for codeine were 2.2, 0.81, and 252 respectively. These were not significantly altered by paracetamol: 2.2, 0.84, and 291 respectively. For codeine-6-glucuronide the values were 2.4, 22.0, and 29.7 respectively. These were not significantly different from those after codeine plus paracetamol: 2.4, 21.9, and 39.6. There were no significant differences between the two treatments in the apparent partial clearances (ml.min-1) of codeine to morphine (88 codeine alone, 70 codeine plus paracetamol), to norcodeine (71 codeine alone, 88 codeine plus paracetamol), and to codeine-6-glucuronide (820 codeine alone, 1022 codeine plus paracetamol). The urinary excretion of codeine-6-glucuronide, morphine, norcodeine, and codeine were not significantly different between the two treatments.
Product Description
Supelco
Codeine-6-β-D-glucuronide solution, 100 μg/mL in methanol: water (2:8), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Norcodeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®