MilliporeSigma
Anti-inflammatory activity of mangostins from Garcinia mangostana.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-22)
Lih-Geeng Chen, Ling-Ling Yang, Ching-Chiung Wang
ABSTRACT
The fruit hull of Garcinia mangostana Linn (Guttiferae) is used as an anti-inflammatory drug in Southeast Asia. Two xanthones, alpha- and gamma-mangostins, were isolated from the fruit hull of G. mangostana, and both significantly inhibited nitric oxide (NO) and PGE(2) production from lipopolysaccharide (LPS)-stimulated RAW 264.7 cells. The IC(50) values for the inhibition of NO production by alpha- and gamma-mangostins were 12.4 and 10.1 microM, respectively. After iNOS enzyme activity was stimulated by LPS for 12 h, treatment with either alpha- or gamma-mangostin at 5 microg/ml (12.2 and 12.6 microM, respectively) for 24 h did not significantly inhibit NO production. The data show that the inhibitory activities of alpha- and gamma-mangostins are not due to direct inhibition of iNOS enzyme activity. On the other hand, expression of iNOS was inhibited by alpha- and gamma-mangostins in LPS-stimulated RAW 264.7 cells, but not by COX-2. However, the level of PGE(2) production was reduced by the two xanthones. In an in vivo study, alpha-mangostin significantly inhibited mice carrageenan-induced paw edema. In conclusion, alpha- and gamma-mangostins from G. mangostana are bioactive substances with anti-inflammatory effects.
Sigma-Aldrich
gamma-Mangostin, ≥98% (HPLC)