MilliporeSigma
  • Separation and determination of five major opium alkaloids with mixed mode of hydrophilic/cation-exchange monolith by pressurized capillary electrochromatography.
Separation and determination of five major opium alkaloids with mixed mode of hydrophilic/cation-exchange monolith by pressurized capillary electrochromatography.
Journal of separation science (2007-10-27)
Xucong Lin, Jiabin Wang, Liyu Li, Xiaochun Wang, Haixia Lü, Zenghong Xie
ABSTRACT
A method for the separation and determination of five major opium alkaloids (narcotine, papaverine, thebaine, codeine, and morphine) in pericarpium papaveris by pressurized CEC (pCEC) with monolithic column has been developed. Under the optimum condition, linear calibration ranges of narcotine, papaverine, thebaine, codeine, and morphine were obtained as 2-85, 2-85, 5-75, 10-65, and 10-65 microg/mL, respectively. LODs of these analytes were 1.5-6.0 microg/mL. The RSD (n=7) of the migration time and peak area were 1.94-5.24 and 4.05-8.21%, respectively. The proposed method was successfully applied to the analysis of pericarpium papaveris samples. Average recoveries of 79.0-95.9% at different fortified levels of alkaloids were achieved with RSD less than 4.6%. Meanwhile, the mechanism of the separation of the alkaloids on the monolithic column was also discussed. The result showed that the separation of alkaloids was mainly based on the mixed mode of hydrophilic interaction (HI) and cation exchange.