MilliporeSigma
  • Enantiomeric deoxycholic acid: total synthesis, characterization, and preliminary toxicity toward colon cancer cell lines.
Enantiomeric deoxycholic acid: total synthesis, characterization, and preliminary toxicity toward colon cancer cell lines.
The Journal of organic chemistry (2007-10-26)
Bryson W Katona, Nigam P Rath, Shrikant Anant, William F Stenson, Douglas F Covey
ABSTRACT
Deoxycholic acid (DCA) is an endogenous secondary bile acid implicated in numerous pathological conditions including colon cancer formation and progression and cholestatic liver disease. DCA involvement in these disease processes results partly from its ability to modulate signaling cascades within the cell, presumably through both direct receptor activation and general detergent mediated membrane changes. To further explore DCA induced changes in cell signaling, we completed a total synthesis of enantiomeric deoxycholic acid (ent-DCA) from achiral 2-methyl-1,3-cyclopentanedione. Using a modified method of the synthesis of ent-testosterone that proceeds through the (R)-(-)-Hajos-Parrish ketone, we have completed the successful synthesis of ent-DCA in 25 steps with a yield of 0.3% with all stereochemical assignments of the product confirmed by X-ray crystallography. Our studies toward this synthesis also uncovered the methodology for the development of a novel A,B-cis steroidal skeleton system containing a C3-C9 single bond as well as conditions to selectively ketalize the typically less reactive 12-carbonyl in poly-keto A,B-cis androgens. The critical micelle concentration (cmc) of ent-DCA, determined by a dye solubilization method, was identical to the cmc of natural DCA. Toxicity studies toward HT-29 and HCT-116 human colon cancer cell lines demonstrated that ent-DCA had similar effects on proliferation, yet showed a markedly decreased ability to induce apoptosis as compared to natural DCA.
Sigma-Aldrich
3-Methyl-1,2-cyclopentanedione, 99%
Sigma-Aldrich
2-Methyl-1,3-cyclopentanedione, 99%
Sigma-Aldrich
Methyl cyclopentenolone, anhydrous, 98%, FCC, FG
Sigma-Aldrich
Methyl cyclopentenolone, anhydrous, natural, 97%, FG
Supelco
Methyl cyclopentenolone, analytical standard