MilliporeSigma
Lithium toxicity in two dogs.
Journal of the South African Veterinary Association (1991-09-01)
N L Davies
ABSTRACT
Two cases of lithium toxicity are reported on in dogs having had lithium hypochlorite chlorinated water as their sole source of drinking water. Clinical signs in one dog included polyuria, polydipsia, loss of body mass; dehydration, diarrhoea and general weakness and in the other case, polyuria, polydipsia, loss of body mass and seizures. Withdrawal of the water resulted in complete recovery.