MilliporeSigma
  • QT Interval prolongation and torsades de pointes due to a coadministration of ciprofloxacin and azimilide in a patient with implantable cardioverter-defibrillator.
QT Interval prolongation and torsades de pointes due to a coadministration of ciprofloxacin and azimilide in a patient with implantable cardioverter-defibrillator.
Pacing and clinical electrophysiology : PACE (2007-08-03)
Jarosław Kaźmierczak, Małgorzata Peregud-Pogorzelska, Ryszard Rzeuski
ABSTRACT
The presented case report describes a male patient with an implanted cardioverter-defibrillator (ICD) in whom a coadministration of ciprofloxacin and azimilide caused QT interval prolongation and multiple episodes of torsades de pointes (TdP) followed by ICD shocks (arrhythmic storm). The case highlights a not described drug interaction between azimilide and ciprofloxacin, which is believed to be the safest member of fluoroquinolones class.
Sigma-Aldrich
Azimilide dihydrochloride, ≥97% (HPLC)