MilliporeSigma
  • Synthesis of (+/-)-3H-epivincamine via a Rh(II)-triggered cyclization/cycloaddition cascade.
Synthesis of (+/-)-3H-epivincamine via a Rh(II)-triggered cyclization/cycloaddition cascade.
Organic letters (2007-07-31)
Dylan B England, Albert Padwa
ABSTRACT
A synthesis of (+/-)-3H-epivincamine is reported. Important steps include (1) a Rh(II)-catalyzed intramolecular [3+2]-cycloaddition of an alpha-diazo indolo amide, (2) a reductive ring opening of the cycloadduct, (3) a decarboethoxylation reaction, and (4) a base-induced keto-amide ring contraction.