MilliporeSigma
  • Infrared spectroscopy of cationized lysine and epsilon-N-methyllysine in the gas phase: effects of alkali-metal ion size and proton affinity on zwitterion stability.
Infrared spectroscopy of cationized lysine and epsilon-N-methyllysine in the gas phase: effects of alkali-metal ion size and proton affinity on zwitterion stability.
The journal of physical chemistry. A (2007-07-20)
Matthew F Bush, Matthew W Forbes, Rebecca A Jockusch, Jos Oomens, Nick C Polfer, Richard J Saykally, Evan R Williams
ABSTRACT
The gas-phase structures of protonated and alkali-metal-cationized lysine (Lys) and epsilon-N-methyllysine (Lys(Me)) are investigated using infrared multiple photon dissociation (IRMPD) spectroscopy utilizing light generated by a free electron laser, in conjunction with ab initio calculations. IRMPD spectra of Lys.Li(+) and Lys.Na(+) are similar, but the spectrum for Lys.K(+) is different, indicating that the structure of lysine in these complexes depends on the metal ion size. The carbonyl stretch of a carboxylic acid group is clearly observed in each of these spectra, indicating that lysine is nonzwitterionic in these complexes. A detailed comparison of these spectra to those calculated for candidate low-energy structures indicates that the bonding motif for the metal ion changes from tricoordinated for Li and Na to dicoordinated for K, clearly revealing the increased importance of hydrogen-bonding relative to metal ion solvation with increasing metal ion size. Spectra for Lys(Me).M(+) show that Lys(Me), an analogue of lysine whose side chain contains a secondary amine, is nonzwitterionic with Li and zwitterionic with K and both forms are present for Na. The proton affinity of Lys(Me) is 16 kJ/mol higher than that of Lys; the higher proton affinity of a secondary amine can result in its preferential protonation and stabilization of the zwitterionic form.
Sigma-Aldrich
Nε-Methyl-L-lysine hydrochloride, ≥98.0% (TLC)