MilliporeSigma
  • Distribution and analgesia of [3H][D-Pen2, D-Pen5]enkephalin and two halogenated analogs after intravenous administration.
Distribution and analgesia of [3H][D-Pen2, D-Pen5]enkephalin and two halogenated analogs after intravenous administration.
The Journal of pharmacology and experimental therapeutics (1991-12-01)
S J Weber, D L Greene, S D Sharma, H I Yamamura, T H Kramer, T F Burks, V J Hruby, L B Hersh, T P Davis
ABSTRACT
To improve pharmacological characteristics of the delta-selective, cyclic peptide [D-Pen2, D-Pen5]enkephalin (DPDPE), modification by halogenation at the Phe4 residue was undertaken. The present study was to determine the extent [3H]DPDPE, [3H][p-Cl-Phe4]DPDPE and [p-125IPhe4]DPDPE crosses the blood-brain barrier, elicits analgesia and to characterize selective organ distribution and stability after i.v. administration. A significantly greater percentage of total [3H][p-Cl-Phe4]DPDPE reached the brain after 10, 20 and 40 min as compared to [3H]DPDPE and both peptides were significantly displaced by pretreatment with naloxone or naltrindole. The amount of [3H]DPDPE detected in the brain was greater than that of [p-125IPhe4]DPDPE. Distribution results revealed large amounts of the administered peptides were sequestered rapidly in the gall bladder and secreted into the small intestine. Hot-plate antinociception tests 5 min after i.v. administration (30 and 60 mg/kg) revealed [p-Cl-Phe4]DPDPE to elicit a much greater analgesic effect as compared to DPDPE or [p-125IPhe4]DPDPE. These results provide evidence that [p-Cl-Phe4]DPDPE has a greater apparent distribution to the brain and has a greater effect on the antinociception threshold as tested on the hot-plate than DPDPE or [p-125IPhe4]DPDPE. Stability of unlabeled and tritiated DPDPE and [p-Cl-Phe4]DPDPE was determined both in vitro and in vivo; both unlabeled and tritiated DPDPE and [p-Cl-Phe4]DPDPE remain intact.