MilliporeSigma
Synthesis of enzyme-degradable, peptide-cross-linked dextran hydrogels.
Bioconjugate chemistry (2007-04-04)
Stéphane G Lévesque, Molly S Shoichet
ABSTRACT
Hydrogels derived from synthetic polymers have been previously engineered to degrade under the activity of matrix metalloproteinases (MMPs). It is believed that these systems can act as extracellular-matrix (ECM) equivalents mimicking the degradation and remodeling of the ECM through the activity of cell-secreted enzymes. In this study, MMP-sensitive hydrogels derived from dextran were developed. In order to avoid the incorporation of hydrolyzable esters often introduced in dextran modification strategies, the polysaccharide was modified with p-maleimidophenyl isocyanate (PMPI) thereby introducing maleimide functionalities in the backbone and resulting in dextran derivatized with p-maleimidophenyl isocyanate (Dex-PMPI). This strategy was favored to separate out the effects of random hydrolysis and enzymatic digestion in the degradation of the dextran hydrogels. A peptide cross-linker, derived from collagen and susceptible to gelatinase A (MMP-2) digestion, was synthesized with bifunctional cysteine termini and used to cross-link the Dex-PMPI. These hydrogels were found to be hydrolytically stable for more than 200 days yet degraded either within 30 h when exposed to bacterial collagenase or within 16 days when exposed to human MMP-2, demonstrating enzymatic-mediated digestion of the peptide cross-links. Further modification of the cross-linked hydrogels with laminin-derived peptides enhanced cell adhesion and survival, demonstrating the potential of these materials for use in tissue engineering applications.
Sigma-Aldrich
4-(Maleinimido)phenyl isocyanate, purum, ≥97.0% (CHN)