MilliporeSigma
  • 1-Methyl-tetrahydro-beta-carboline-3-carboxylic acid is present in the rat brain and is not increased after acute ethanol injection with cyanamide treatment.
1-Methyl-tetrahydro-beta-carboline-3-carboxylic acid is present in the rat brain and is not increased after acute ethanol injection with cyanamide treatment.
Alcohol (Fayetteville, N.Y.) (1992-01-01)
S Fukushima, K Matsubara, A Akane, H Shiono
ABSTRACT
We conducted analyses of 1-methyl-1,2,3,4-tetrahydro-beta-carboline-3-carboxylic acid (1Me3C-THBC) by gas chromatography-mass spectrometry (negative chemical ionization mode) to investigate its presence and the in vivo condensation between tryptophan and AcH. 1Me3C-THBC was found in the cerebellum and the cerebrum of normal rat [117.0 +/- 41.7 and 46.5 +/- 13.9 pmol/g tissue (mean +/- SEM), respectively]. The concentrations of 1Me3C-THBC and tryptophan were higher in the cerebellum than those in the cerebrum. The level of 1Me3C-THBC in both regions remained unchanged following a single oral ethanol administration alone or with cyanamide pretreatment. These data suggest that acetaldehyde is an unlike precursor of 1Me3C-THBC as a result of ethanol ingestion. 1Me3C-THBC also existed in the rat chow (282.0 +/- 24.2 pmol/g), so that most of brain 1Me3C-THBC detected in the rat brain might have originated from dietary sources. However, the possibility of a biosynthesis from tryptophan and alpha-keto acid still remained, especially after long-term ethanol treatment.
Sigma-Aldrich
1,2,3,4-Tetrahydroharmane-3-carboxylic acid