MilliporeSigma
Mutasynthesis - uniting chemistry and genetics for drug discovery.
Trends in biotechnology (2007-02-20)
Kira J Weissman
ABSTRACT
Mutasynthesis couples the power of chemical synthesis with molecular biology to generate derivatives of medicinally valuable, natural products. Recently, this technique has been exploited by Cambridge-based biotech company Biotica Technology Ltd, and their collaborators, to generate promising new variants of the polyketide anti-cancer compounds rapamycin and borrelidin.
Sigma-Aldrich
Borrelidin, from Streptomyces parvulus, ≥98% (HPLC)