MilliporeSigma
[Studies on chemical constituents in herb from Artemisia rupestris].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2007-01-31)
Wei-Xia Song, Teng-Fei Ji, Yi-Kang Si, Ya-Lun Su
ABSTRACT
To study the chemical constituents of Artemisia rupestris. The chemical constituents were isolated by column chromatography on silical gel and sephadex LH - 20. Their structures were elucidated on the basis of spectral analysis. 8 compounds have isolated from this plant, and the structures of them have identified as rupestonic acid (1), chrysosplenetin B (2), artemetin (3), herniarin (4), isokaempferide (5), vanillic acid (6), kaempferol 3, 3', 4'-trimethyl ether (7) and ermanine (8). Compounds 2-8 have been isolated from this plant for the first time.
Sigma-Aldrich
7-Methoxycoumarin, ≥98%
Sigma-Aldrich
7-Methoxycoumarin, suitable for fluorescence, ≥98.0% (TLC)