MilliporeSigma
  • Protective effects of ginsenoside Rg2 against glutamate-induced neurotoxicity in PC12 cells.
Protective effects of ginsenoside Rg2 against glutamate-induced neurotoxicity in PC12 cells.
Journal of ethnopharmacology (2007-01-30)
Na Li, Bin Liu, Dean E Dluzen, Yi Jin
ABSTRACT
We investigated the effect of ginsenoside Rg2 on neurotoxic activities induced by glutamate in PC12 cells. The cells were incubated with glutamate (1 mmol/L), glutamate and ginsenoside Rg2 (0.05, 0.1, 0.2 mmol/L) or nimodipine (5 micromol/L) for 24 h. The cellular viability was assessed by MTT assay. The lipid peroxidation products malondialdehyde (MDA) and nitrogen oxide (NO) were measured by a spectrophotometric method. Fura-2/AM, as a cell permeable fluorescent probe for Ca2+, was used to detect intracellular Ca2+ concentration ([Ca2+]i) using a monespectrofluorometer. Immunocytochemical techniques were employed to check the protein expression levels of calpain II, caspase-3 and beta-amyloid (Abeta)1-40 in PC12 cells. The results showed that glutamate decreased the cell viability, increased [Ca2+]i, lipid peroxidation (the excessive production of MDA, NO) and the protein expression levels of calpain II, caspase-3 and Abeta1-40 in PC12 cells. Ginsenoside Rg2 significantly attenuated glutamate-induced neurotoxic effects upon these parameters at all doses tested. Our study suggests that ginsenoside Rg2 has a neuroprotective effect against glutamate-induced neurotoxicity through mechanisms related to anti-oxidation and anti-apoptosis. In addition, the inhibitory effect of ginsenoside Rg2 against the formation of Abeta1-40 suggests that ginsenoside Rg2 may also represent a potential treatment strategy for Alzheimer's disease.
Product Description
Supelco
Ginsenoside Rg2, analytical standard