MilliporeSigma
  • In vivo selection for the enhancement of Thermotoga maritima exopolygalacturonase activity at neutral pH and low temperature.
In vivo selection for the enhancement of Thermotoga maritima exopolygalacturonase activity at neutral pH and low temperature.
Protein engineering, design & selection : PEDS (2007-01-16)
Jullien Drone, Michel Dion, Charles Tellier, Claude Rabiller
ABSTRACT
The aim of this study was to develop an Escherichia coli-based metabolic selection system for the uncovering of new oligogalacturonate-active enzymes. Based on the expression of the specific permease TogMNAB, this system enabled the entry of oligogalacturonates into the cytoplasm of E. coli thus providing a modified strain usable for this purpose. This tool was used for the metabolic selection of Thermotoga maritima exopolygalacturonase (TmGalU) mutants enabling the uptake of sodium trigalacturonate as the sole carbon source by the bacterium. In only one round of error-prone PCR and selection, mutants of TmGalU with a 4-fold increased turnover at pH 7.0 and 2-fold more active at 37 degrees C than wild-type enzyme were isolated. These results show the versatility of this strain for the evolution of oligogalacturonate-active enzymes.
Sigma-Aldrich
Trigalacturonic acid, ≥90% (HPLC)