MilliporeSigma
  • Interactions between buprenorphine and antiretrovirals. I. The nonnucleoside reverse-transcriptase inhibitors efavirenz and delavirdine.
Interactions between buprenorphine and antiretrovirals. I. The nonnucleoside reverse-transcriptase inhibitors efavirenz and delavirdine.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (2006-11-17)
Elinore F McCance-Katz, David E Moody, Gene D Morse, Gerald Friedland, Patricia Pade, Jennifer Baker, Anika Alvanzo, Patrick Smith, Abayomi Ogundele, Peter Jatlow, Petrie M Rainey
ABSTRACT
This study examined drug interactions between buprenorphine, an opioid partial agonist medication used in the treatment of opioid dependence, and the nonnucleoside reverse-transcriptase inhibitors (NNRTIs) efavirenz (EFV) and delavirdine (DLV). Opioid-dependent, buprenorphine/naloxone-maintained, human immunodeficiency virus (HIV)-negative volunteers (n=10 per NNRTI) participated in 24-h sessions to determine pharmacokinetics of buprenorphine and of buprenorphine with either EFV or DLV after administration of standard doses of either antiretroviral for 15 or 7 days, respectively. Opiate withdrawal symptoms, cognitive effects, and adverse events were determined before and after antiretroviral administration in opioid-dependent participants. The pharmacokinetics of NNRTIs in healthy control participants were used to determine the effect of buprenorphine on NNRTIs. EFV decreased the buprenorphine area under the concentration-time curve (P<.001). DLV increased buprenorphine concentrations (P<.001). Clinically significant consequences of these interactions were not observed. Buprenorphine did not alter antiretroviral pharmacokinetics. Adjustments of doses of either buprenorphine or EFV or DLV are not likely to be necessary when these drugs are administered for the treatment of opiate dependence and HIV disease.
Sigma-Aldrich
Delavirdine mesylate, ≥98% (HPLC)