MilliporeSigma
  • Purification of human malaria parasite hypoxanthine guanine xanthine phosphoribosyltransferase (HGXPRT) using immobilized Reactive Red 120.
Purification of human malaria parasite hypoxanthine guanine xanthine phosphoribosyltransferase (HGXPRT) using immobilized Reactive Red 120.
Protein expression and purification (2006-11-14)
Boniface Mbewe, Kelly Chibale, David B McIntosh
ABSTRACT
Malaria is caused by Plasmodium parasite infection. The human malarial parasite does not have a de novo pathway for synthesis of nucleotides and the purine salvage pathway enzyme hypoxanthine guanine xanthine phosphoribosyltransferase (HGXPRT) is critical for survival. In our efforts to find inhibitors of the malarial parasite HGXPRT, we have developed a simple but effective purification protocol for this protein expressed in Escherichia coli without an affinity tag. The protocol consists of tandem columns of anion exchange and immobilized Reactive Red 120 resins. The enzyme is inactive as isolated but can be activated by incubation with substrate(s).
Sigma-Aldrich
Reactive Red 120