MilliporeSigma
  • Vibrational spectra and ab initio analysis of tert-butyl, trimethylsilyl, trimethylgermyl, trimethylstannyl, and trimethylplumbyl derivatives of 3,3-dimethylcyclopropene IX. 3,3-Dimethyl-1-(trimethylplumbyl)cyclopropene.
Vibrational spectra and ab initio analysis of tert-butyl, trimethylsilyl, trimethylgermyl, trimethylstannyl, and trimethylplumbyl derivatives of 3,3-dimethylcyclopropene IX. 3,3-Dimethyl-1-(trimethylplumbyl)cyclopropene.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-11-10)
G R De Maré, Yu N Panchenko, A V Abramenkov
ABSTRACT
The geometrical parameters and quantum mechanical force fields (QMFF's) of 3,3-dimethyl-1-(trimethylplumbyl)cyclopropene (I), 3,3-dimethyl-1-(t-butyl)cyclopropene (II), 3,3-dimethyl-1-(trimethylsilyl)cyclopropene (III), 3,3-dimethyl-1-(trimethylgermyl)cyclopropene (IV), and 3,3-dimethyl-1-(trimethylstannyl)cyclopropene (V) were calculated at the pseudopotential (HF/SDDAll) level. Analysis of the optimised geometrical parameters was performed. The set of scale factors for correction of the pseudopotential QMFF of III was determined using its earlier well-characterised vibrational spectrum. Transferral of the set of scale factors obtained for III to the QMFF's of I, II, IV and V was followed by calculation of the fundamental vibrational frequencies. Analysis of the results for these molecules revealed some peculiarities in the vibrational frequencies obtained at the pseudopotential level.