MilliporeSigma
  • Further studies on imidazo[4,5-b]pyridine AT1 angiotensin II receptor antagonists. Effects of the transformation of the 4-phenylquinoline backbone into 4-phenylisoquinolinone or 1-phenylindene scaffolds.
Further studies on imidazo[4,5-b]pyridine AT1 angiotensin II receptor antagonists. Effects of the transformation of the 4-phenylquinoline backbone into 4-phenylisoquinolinone or 1-phenylindene scaffolds.
Journal of medicinal chemistry (2006-10-27)
Andrea Cappelli, Gal la Pericot Mohr, Germano Giuliani, Simone Galeazzi, Maurizio Anzini, Laura Mennuni, Flora Ferrari, Francesco Makovec, Eva M Kleinrath, Thierry Langer, Massimo Valoti, Gianluca Giorgi, Salvatore Vomero
ABSTRACT
The 4-phenylquinoline fragment of novel AT(1) receptor antagonists 4 based on imidazo[4,5-b]pyridine moiety was replaced by 4-phenylisoquinolinone (compounds 5) or 1-phenylindene (compounds 6) scaffolds to investigate the structure-activity relationships. Binding studies showed that most of the synthesized compounds display high affinity for the AT(1) receptor. Because of the in vitro high potency of carboxylic acids 5b,f, they were evaluated in permeability (in Caco-2 cells) and in pharmacokinetic studies in comparison with quinoline derivatives 4b,i,j,k. The studies showed that these compounds are characterized by rapid excretion, low membrane permeability, and low oral bioavailability. The structure optimization of the indene derivatives led to compounds 6e,f possessing interesting AT(1) receptor affinities. Optimization produced polymerizing AT(1) receptor ligand 6c, which forms a thermoreversible polymer (poly-6c) and is released from the latter by a temperature-dependent kinetics. The results suggest the possibility of developing novel polymeric prodrugs based on a new release mechanism. Finally, a set of 34 AT(1) receptor antagonists was used as a new test for the evaluation of the predictive capability of the previously published qualitative and quantitative pharmacophore models.
Sigma-Aldrich
4-Azabenzimidazole, 99%