MilliporeSigma
  • Determination of benzimidazole fungicides in soil samples using microwave-assisted micellar extraction and liquid chromatography with fluorescence detection.
Determination of benzimidazole fungicides in soil samples using microwave-assisted micellar extraction and liquid chromatography with fluorescence detection.
Journal of AOAC International (2006-10-18)
Radoslav Halko, Carolina Padrón Sanz, Zoraida Sosa Ferrera, José Juan Santana Rodríguez
ABSTRACT
A simple and fast analytical method was developed for the determination of benzimidazole fungicides (benomyl, carbendazim, thiabendazole, and fuberidazole) in soil samples. The analytes were extracted from the soil samples by means of conventional microwave-assisted extraction, using the non-ionic surfactants polyoxyethylene 10 lauryl ether (POLE) and oligoethylene glycol monooalkyl ether (Genapol X-080) as extractants. Determinations were made by using liquid chromatography with direct fluorescence detection. The use of an analytical column Symmetry C-18 offered short retention times of analytes without the need of any pH regulators with mobile phase methanol-water (50 + 50, v/v). The best results were obtained using 5% (v/v) POLE as extractant with recoveries of the fungicides in spiked soil samples between 71 and 105%. The results were compared with those obtained when Soxhlet extraction was applied to the same soil samples.
Sigma-Aldrich
Genapol® X-100
Sigma-Aldrich
Genapol® X-080