MilliporeSigma
  • Verification of cefmetazole and cefpodoxime proxetil contamination to other pharmaceuticals by liquid chromatography-tandem mass spectrometry.
Verification of cefmetazole and cefpodoxime proxetil contamination to other pharmaceuticals by liquid chromatography-tandem mass spectrometry.
Chemical & pharmaceutical bulletin (2006-10-04)
Naoto Fukutsu, Yuich Sakamaki, Takao Kawasaki, Koichi Saito, Hiroyuki Nakazawa
ABSTRACT
Cross-contamination is a critical issue for pharmaceutical manufacturing, especially for beta-lactam antibiotics. Thus, an analytical method for the simultaneous determination of beta-lactam antibiotics cefmetazole (CMZ) and cefpodoxime proxetil (CPDXPR) contaminants in non-beta-lactam pharmaceuticals was developed using high-performance liquid chromatography-tandem mass spectrometry. The developed method was found to be sensitive at the detection limit of 0.002 ppm for both compounds. Mean recoveries of CMZ and CPDXPR from olmesartan medoxomil (OLM) tablets were 96.7 to 102.2% and 88.9 to 94.2%, respectively. The developed method was successfully applied for the verification of CMZ and CPDXPR contamination to actually manufactured OLM tablets.
Sigma-Aldrich
Cefmetazole sodium salt
USP
Cefpodoxime proxetil, United States Pharmacopeia (USP) Reference Standard
Cefpodoxime proxetil, European Pharmacopoeia (EP) Reference Standard
Cefpodoxime proxetil for impurity H identification, European Pharmacopoeia (EP) Reference Standard
Cefpodoxime proxetil for peak identification, European Pharmacopoeia (EP) Reference Standard