MilliporeSigma
  • The dopamine agonist piribedil with L-DOPA improves attentional dysfunction: relevance for Parkinson's disease.
The dopamine agonist piribedil with L-DOPA improves attentional dysfunction: relevance for Parkinson's disease.
The Journal of pharmacology and experimental therapeutics (2006-08-22)
Nathalie Turle-Lorenzo, Béatrice Maurin, Carole Puma, Claire Chezaubernard, Philippe Morain, Christelle Baunez, André Nieoullon, Marianne Amalric
ABSTRACT
Cognitive deficits are often associated with motor symptoms in Parkinson's disease. This study investigates the ability of piribedil ([(methylenedioxy-3,4 benzyl)-4 pyperazinyl-1]-2 pyrimidine), a D(2)/D(3) dopamine (DA) receptor agonist with antagonist activity at alpha(2A)-adrenoceptors, to restore motor and attentional deficits in nigrostriatal 6-hydroxydopamine-lesioned rats. Subjects were trained to depress a lever, detect a stimulus occurring after variable foreperiods, and release the lever quickly afterward. Striatal DA depletions produce deficits in the timing of foreperiods and prolong reaction times. Although a subchronic treatment with piribedil (0.1-2 mg/kg) is not effective, a dose of 0.3 mg/kg administered for 3 weeks significantly reverses the akinetic deficits produced by the striatal dopamine depletion and progressively improves attentional deficits. When coadministered with the dopamine prodrug l-3,4-dihydroxyphenylalanine (l-DOPA) (3 mg/kg), piribedil (0.3 mg/kg) promotes a rapid and full recovery of preoperative performance. These results suggest that administration of l-DOPA in combination with piribedil in a chronic treatment as either initial or supplemental therapy for Parkinson's disease might improve cognitive functions while reducing the risk for motor complications.