MilliporeSigma
Phenylalkylamine alkaloids from Stapelia hirsuta L.
Natural product research (2006-06-07)
Marwan Shabana, Mariam Gonaid, Maha Mahmoud Salama, Essam Abdel-Sattar
ABSTRACT
Four alkaloids of the phenethylamine derivatives have been isolated from the n-butanol fraction of the aerial parts of Stapelia hirsuta L. The structures of the isolated alkaloids were determined as N-acetyl hordenine (a new natural compound), hordenine, candicine and hordenine-1-O-beta-D-glucoside, in addition to luteolin-7-O-beta-D-glucopyranoside.
Sigma-Aldrich
Hordenine, ≥97.0% (HPLC)