MilliporeSigma
Pharmacological evaluation of halogenated delta 8-THC analogs.
Pharmacology, biochemistry, and behavior (1991-11-01)
A Charalambous, S Lin, G Marciniak, A Banijamali, F L Friend, D R Compton, B R Martin, A Makriyannis
ABSTRACT
-(-)-5'-Bromo-delta 8-THC, (-)-5'-trifluoromethyl-delta 8-THC, (-)-5'-iodo-delta 8-THC, (-)-5'-fluoro-delta 8-THC, (-)-11-fluoro-delta 8-THC and (-)-2-iodo-delta 8-THC were synthesized and evaluated in male ICR mice for their effects on sedation, temperature, catalepsy and antinociception following intravenous injection. The analogs were also tested for relative affinities for cannabinoid binding sites derived from rat cortex membranes, using [3H] CP-55,940 as the tritiated ligand. The results showed that the 5'-bromo, 5'-iodo and 5'-trifluoromethyl analogs were 2-40 times more potent than (-)-delta 8-THC in all biological tests, while the 5'-fluoro and 11-fluoro derivatives were less active. With the 2-iodo analog, a 12-fold separation was observed between antinociception and sedation, pointing to the importance of the side chain orientation in determining cannabinoid activity and to the possible involvement of more than one cannabinoid receptor site. The pharmacological data closely paralleled the data obtained from the binding assay.
Product Description
Supelco
(−)-Δ8-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®