MilliporeSigma
  • Simultaneous determination of zonisamide, carbamazepine and carbamazepine-10,11-epoxide in infant serum by high-performance liquid chromatography.
Simultaneous determination of zonisamide, carbamazepine and carbamazepine-10,11-epoxide in infant serum by high-performance liquid chromatography.
Journal of pharmaceutical and biomedical analysis (2006-04-07)
Terumitsu Yoshida, Koichi Imai, Sumiko Motohashi, Shin-ichiro Hamano, Masato Sato
ABSTRACT
This study developed a simple method for the simultaneous determination of zonisamide (ZNS), carbamazepine (CBZ) and its active metabolite, carbamazepine-10,11-epoxide (CBZE) in infant serum using reversed-phase high-performance liquid chromatograph (HPLC). The method involves a single-step protein precipitation procedure that uses no solid-phase or liquid-liquid extraction. The HPLC separation was carried out on a Cadenza CD-C18 column (3 microm, 4.6 mm x 150 mm) with potassium phosphate buffer (pH 4.6; 25 mM)-methanol-acetonitrile (65:20:15 (v/v/v)) as a mobile phase at a 1.0 ml/min flow rate: ZNS was detectable using a UV detector at 235 nm, and both CBZ and CBZE were at 215 nm. The quantification limits were established in accordance with each therapeutic range at 2.5 microg/ml for ZNS, 0.5 microg/ml for CBZ, and 0.25 microg/ml for CBZE. The respective coefficients of variation were 1.3-6.0% and 2.2-7.7% for the intra- and inter-assay.
Product Description
Supelco
Carbamazepine 10,11-epoxide, analytical standard
Supelco
Carbamazepine-10,11-epoxide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®