MilliporeSigma
  • Two new species of Epistylis (Ciliophora: Peritrichida) on the blue crab (Callinectes sapidus) in the Gulf of Mexico.
Two new species of Epistylis (Ciliophora: Peritrichida) on the blue crab (Callinectes sapidus) in the Gulf of Mexico.
The Journal of eukaryotic microbiology (2006-04-04)
Hongwei Ma, Robin M Overstreet
ABSTRACT
Two epibiotic peritrichs infested the blue crab, Callinectes sapidus, from the Gulf of Mexico, Mississippi, USA. Epistylis callinectes n. sp. was isolated from the epipods of maxillipeds, bases of gill-cleaning setae, and gills, and Epistylis clampi n. sp. was isolated from the exterior surfaces of the exoskeleton. Epistylis callinectes has short, symmetrically and dichotomously branched stalks; its zooid is elongate ovoid and conspicuously longer than the individual stalk branches, measuring 40-57 (49) x 18-33 (26) microm in vivo and containing a thick, undivided peristomial lip (PL). It has a single contractile vacuole and a transverse horseshoe-shaped macronucleus. Its haplokinety (H) and polykinety (Po) complete one and one-half circuits on the peristome before entering the infundibulum. There is a distal kinetal fragment present at the distal end of both the H and Po. Epistylis callinectes has 48-70 transverse silverlines from the oral area to the trochal band (TB) and 19-26 from the TB to the scopula. Epistylis clampi has long, asymmetrically, and dichotomously branched stalks. Its zooid is elongate vase-shaped, measuring 35-64 (48) x 21-30 (27) microm in vivo and with a thick, transversely folded PL. The stalks supporting zooids are unequal in length. Its zooid has a single contractile vacuole and a transverse horseshoe-shaped macronucleus occurs in the upper half of the body. Its H and Po complete approximately one circuit around the peristome before entering the infundibulum. There is a distal kinetal fragment present at the distal end of both the H and Po. This species has 71-112 transverse silverlines from the peristome to the scopula.
Sigma-Aldrich
Silver proteinate, ~8% Ag basis