MilliporeSigma
  • A preliminary neutron diffraction study of rasburicase, a recombinant urate oxidase enzyme, complexed with 8-azaxanthin.
A preliminary neutron diffraction study of rasburicase, a recombinant urate oxidase enzyme, complexed with 8-azaxanthin.
Acta crystallographica. Section F, Structural biology and crystallization communications (2006-03-03)
Monika Budayova-Spano, Françoise Bonneté, Natalie Ferté, Mohamed El Hajji, Flora Meilleur, Matthew Paul Blakeley, Bertrand Castro
ABSTRACT
Crystallization and preliminary neutron diffraction measurements of rasburicase, a recombinant urate oxidase enzyme expressed by a genetically modified Saccharomyces cerevisiae strain, complexed with a purine-type inhibitor (8-azaxanthin) are reported. Neutron Laue diffraction data were collected to 2.1 A resolution using the LADI instrument from a crystal (grown in D2O) with volume 1.8 mm3. The aim of this neutron diffraction study is to determine the protonation states of the inhibitor and residues within the active site. This will lead to improved comprehension of the enzymatic mechanism of this important enzyme, which is used as a protein drug to reduce toxic uric acid accumulation during chemotherapy. This paper illustrates the high quality of the neutron diffraction data collected, which are suitable for high-resolution structural analysis. In comparison with other neutron protein crystallography studies to date in which a hydrogenated protein has been used, the volume of the crystal was relatively small and yet the data still extend to high resolution. Furthermore, urate oxidase has one of the largest primitive unit-cell volumes (space group I222, unit-cell parameters a = 80, b = 96, c = 106 A) and molecular weights (135 kDa for the homotetramer) so far successfully studied with neutrons.
Sigma-Aldrich
8-Azaxanthine monohydrate, ≥98.0% (HPLC)