MilliporeSigma
  • Simultaneous determination of nine saponins from Panax notoginseng using HPLC and pressurized liquid extraction.
Simultaneous determination of nine saponins from Panax notoginseng using HPLC and pressurized liquid extraction.
Journal of pharmaceutical and biomedical analysis (2005-12-01)
J B Wan, C M Lai, S P Li, M Y Lee, L Y Kong, Y T Wang
ABSTRACT
A HPLC and pressurized liquid extraction (PLE) method was developed for simultaneous determination of nine saponins, including notoginsenoside R1, ginsenoside Rg1, Re, Rf, Rb1, Rc, Rb2, Rb3 and Rd in Panax notoginseng. The analysis was performed on C18 column with water-acetonitrile gradient elution and the investigated saponins were authenticated by comparing retention time and mass spectra with their reference compounds. Several methods including PLE, ultrasonication, soxhlet extraction and immersion were used for sample preparation and their extraction efficiency was compared. The results showed that PLE has the highest extraction efficiency and repeatability, which would be valuable on standardization of sample preparation for quality control of Chinese medicines. The developed HPLC and PLE is an effective approach for simultaneously quantitative determination of sapoinins in P. notoginseng, which could be used for quality control of P. notoginseng and its preparations.
Sigma-Aldrich
Notoginsenoside R1, ≥98% (HPLC)