MilliporeSigma
  • Protein kinase A and Ca(v)1 (L-Type) channels are common targets to facilitatory adenosine A2A and muscarinic M1 receptors on rat motoneurons.
Protein kinase A and Ca(v)1 (L-Type) channels are common targets to facilitatory adenosine A2A and muscarinic M1 receptors on rat motoneurons.
Neuro-Signals (2005-11-23)
Laura Oliveira, Paulo Correia-de-Sá
ABSTRACT
At the rat motor endplate, pre-synaptic facilitatory adenosine A2A and muscarinic M1 receptors are mutually exclusive. We investigated whether these receptors share a common intracellular signalling pathway. Suppression of McN-A-343-induced M1 facilitation of [3H]ACh release was partially recovered when CGS21680C (an A2A agonist) was combined with the cyclic AMP antagonist Rp-cAMPS. Forskolin, rolipram and 8-bromo-cyclic AMP mimicked CGS21680C blockade of M1 facilitation. Both Rp-cAMPs and nifedipine reduced augmentation of [3H]ACh release by McN-A-343 and CGS21680C. Activation of M1 and A2A receptors enhanced Ca2+ recruitment through nifedipine-sensitive channels. Nifedipine inhibition revealed by McN-A-343 was prevented by chelerythrine (a PKC inhibitor) and Rp-cAMPS, suggesting that Ca(v)1 (L-type) channels phosphorylation by PKA and PKC is required. Rp-cAMPS inhibited [3H]ACh release in the presence of phorbol 12-myristate 13-acetate, but PKC inhibition by chelerythrine had no effect on release in the presence of 8-bromo-cyclic AMP. This suggests that the involvement of PKA may be secondary to M1-induced PKC activation. In conclusion, competition of M1 and A2A receptors to facilitate ACh release from motoneurons may occur by signal convergence to a common pathway involving PKA activation and Ca2+ influx through Ca(v)1 (L-type) channels.