MilliporeSigma
  • Enhancing effect of hydroxypropyl-beta-cyclodextrin on cutaneous penetration and activation of ethyl 4-biphenylyl acetate in hairless mouse skin.
Enhancing effect of hydroxypropyl-beta-cyclodextrin on cutaneous penetration and activation of ethyl 4-biphenylyl acetate in hairless mouse skin.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2005-11-01)
H Arima, T Miyaji, T Irie, F Hirayama, K Uekama
ABSTRACT
The effect of hydroxypropyl-beta-cyclodextrin (HP-beta-CyD) on the cutaneous penetration and activation of ethyl 4-biphenylyl acetate (EBA), a prodrug of non-steroidal anti-inflammatory drug 4-biphenylylacetic acid (BPAA), from hydrophilic ointment was investigated, using hairless mouse skin in vitro. When the hydrophilic ointment containing a complex of EBA with HP-beta-CyD was applied to the full-thickness skin, HP-beta-CyD facilitated the penetration of EBA into the skin, the conversion of EBA to BPAA in the epidermis and the transfer of BPAA to the receptor phase. Under the present condition, pre- and post-application of the ointment containing HP-beta-CyD onto the skin did not affect the cutaneous penetration of EBA and its activation. When the ointment containing the EBA:HP-beta-CyD complex was applied to the skin, the flux of BPAA through the tape-stripped skin was greater than that through the full-thickness skin, while the activation of the prodrug in the skin was slowed down by the tape-stripping. When propylene glycol was used as a vehicle, HP-beta-CyD no longer enhanced the cutaneous permeation of BPAA through the full-thickness skin. These results suggest that the enhancing effect of HP-beta-CyD on the cutaneous penetration of EBA would be ascribable largely to an increase in effective concentration of EBA in the ointment. Furthermore, the slow diffusion of EBA solubilized in HP-beta-CyD through the stratum corneum, together with the vehicle effect, could make the prodrug more susceptible to the metabolic process that is active in the epidermis, eventually leading to the facilitated activation of the prodrug.
Sigma-Aldrich
4-Biphenylacetic acid, 98%