MilliporeSigma
  • Metaraminol (Aramine) in the management of a significant amlodipine overdose.
Metaraminol (Aramine) in the management of a significant amlodipine overdose.
Human & experimental toxicology (2005-08-27)
D M Wood, K D Wright, A L Jones, P I Dargan
ABSTRACT
To report a patient with a significant amlodipine self-poisoning who failed to clinically respond to conventional treatment and was managed with metaraminol (Aramine). A 43-year old male presenting after ingestion of 560 mg amlodipine, who failed to respond clinically to treatment with fluid resuscitation, calcium salts, glucagon and norepinephrine/epinephrine inotropic support. Following a loading bolus of 2 mg and intravenous infusion (83 microg/min) of metaraminol (Aramine) there was improvement in his blood pressure, cardiac output and urine output. This is the first case report of the beneficial use of metaraminol (aramine) in the management of significant amlodipine poisoning unresponsive to conventional therapy.
Sigma-Aldrich
Metaraminol (+)-bitartrate salt
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
US$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
US$60.60