MilliporeSigma
  • [Study on chemical constituents in herbs of Anoectochilus roxburghii II].
[Study on chemical constituents in herbs of Anoectochilus roxburghii II].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2005-08-04)
Chun-nian He, Chun-lan Wang, Shun-xing Guo, Jun-shan Yang, Pei-gen Xiao
ABSTRACT
To study the chemical constituents of Anoectochilus roxburghii. Silica gel and sephadex LH-20 column chromotography were used in the isolation from the ethanol extracts of the whole plant, the compounds were determined on the basis of various modern spectroscopic analysis and physical constants. Five compounds were isolated from the CHCl3 soluble portion, identified as p-Hydroxybenzaldehyde (I), ferulic acid (II), quercetin (III), daucosterol (IV), cirsilineol (V). All these compounds were isolated from the plant for the first time, compound V was isolated from the Orchid Family for the first time, the other compounds were isolated from this genus for the first time.