MilliporeSigma
  • Intracerebroventricular self-administration of commonly abused anabolic-androgenic steroids in male hamsters (Mesocricetus auratus): nandrolone, drostanolone, oxymetholone, and stanozolol.
Intracerebroventricular self-administration of commonly abused anabolic-androgenic steroids in male hamsters (Mesocricetus auratus): nandrolone, drostanolone, oxymetholone, and stanozolol.
Behavioral neuroscience (2005-07-07)
Cortney L Ballard, Ruth I Wood
ABSTRACT
Previous studies by the authors have shown voluntary intracerebroventricular (icv) testosterone self-administration in hamsters (Mesocricetus auratus). Here, the authors compared icv self-administration of 4 anabolic steroids (drostanolone, nandrolone, oxymetholone, and stanozolol) at 0.1, 1.0, and 2.0 microg/microl, each for 8 days. Males (n=8/group) showed the highest levels of operant behavior for injectable steroids (drostanolone, nandrolone) compared with orally active androgens (oxymetholone, stanozolol). For nandrolone, responses on the active and inactive nose-pokes averaged 22.3 +/- 4.6/4 hr and 10.7 +/- 2.0/4 hr, respectively. Responding for drostanolone was similar. Males self-administering oxymetholone or stanozolol did not prefer the active nose-poke. These data demonstrate that injectable androgens are more reinforcing than oral steroids.