- Fancy bioisosteres: metallocene-derived G-protein-coupled receptor ligands with subnanomolar binding affinity and novel selectivity profiles.
Fancy bioisosteres: metallocene-derived G-protein-coupled receptor ligands with subnanomolar binding affinity and novel selectivity profiles.
Metallocene-derived bioisosteres lead to exceptionally strong binding G-protein-coupled receptor ligands, indicating substantial plasticity of the receptor excluded volume. Novel binding profiles of ferrocenylcarboxamides combining subnanomolar Ki values for the dopamine D4 receptor (1a, 0.52 nM; 1b, 0.63 nM) with superpotent serotonin 5-hydroxytryptamine1A (1a, 0.50 nM) and dopamine D3 receptor binding (1b, 0.64 nM) and selective D4 agonist properties of the ruthenocene 1c may be a starting point for highly beneficial central nervous system active drugs.