MilliporeSigma
  • Targeting the tumor-associated folate receptor with an 111In-DTPA conjugate of pteroic acid.
Targeting the tumor-associated folate receptor with an 111In-DTPA conjugate of pteroic acid.
Journal of the American Chemical Society (2005-05-19)
Chun-Yen Ke, Carla J Mathias, Mark A Green
ABSTRACT
The cell membrane folate receptor is a potential molecular target for tumor-selective drug delivery. To probe structural requirements for folate receptor targeting with low molecular weight radiometal chelates, specifically the role of the amino acid fragment of folic acid (pteroylglutamic acid) in mediating targeting selectivity, the amide-linked conjugate pteroyl-NHCH(2)CH(2)OCH(2)CH(2)OCH(2)CH(2)NH-DTPA was prepared by a three-step procedure from pteroic acid, 2,2'-(ethylenedioxy)-bis(ethylamine), and t-Bu-protected DTPA. This conjugate, 1-{2-[2-[(2-(biscarboxymethyl-amino)ethyl)-carboxymethyl-amino]ethyl]-carboxymethyl-amino}-acetylamino-3,6-dioxa-8-pteroylamino-octane (1), was employed for synthesis of the corresponding (111)In(III) radiopharmaceutical. Following intravenous administration to athymic mice, the (111)In complex of 1 was found to selectively localize in folate receptor-positive human KB tumor xenografts and to afford prolonged tumor retention of the (111)In radiolabel (5.4 +/- 0.8, 5.6 +/- 1.1, and 3.6 +/- 0.6% of the injected dose per gram of tumor at 1, 4, and 24 h, respectively). The observed tumor localization was effectively blocked by co-administration of folic acid with the (111)In-1 complex, consistent with a folate receptor-mediated targeting process. In control studies, tumor targeting with this pteroic acid conjugate appears as effective as that seen using (111)In-DTPA-folate, a radiopharmaceutical that has progressed to clinical trials for detection of folate receptor-expressing gynecological tumors.
Product Description
Folic acid impurity D, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pteroic acid, ≥93%
Sigma-Aldrich
N10-(Trifluoroacetyl)pteroic acid, 95%