MilliporeSigma
  • Alprazolam as a probe for CYP3A using a single blood sample: pharmacokinetics of parent drug, and of alpha- and 4-hydroxy metabolites in healthy subjects.
Alprazolam as a probe for CYP3A using a single blood sample: pharmacokinetics of parent drug, and of alpha- and 4-hydroxy metabolites in healthy subjects.
European journal of clinical pharmacology (2005-04-05)
Agneta Wennerholm, Annika Allqvist, Jan-Olof Svensson, Lars L Gustafsson, Rajaa A Mirghani, Leif Bertilsson
ABSTRACT
(1) To determine the pharmacokinetic parameters of alprazolam and its two metabolites in plasma from healthy volunteers; (2) to identify a suitable single time point to take a plasma sample for CYP3A phenotyping. Twelve healthy Swedish volunteers received a single oral dose of 1 mg alprazolam. Blood samples were collected before drug intake and frequently up to 72 h thereafter. A liquid-chromatography/mass-spectrometry (LC/MS) method was used for the quantification of alprazolam, and 4- and alpha-hydroxyalprazolam. The interindividual variation in the area under the concentration-time curve (AUC) was two, three and fourfold for alprazolam, 4-hydroxyalprazolam and alpha-hydroxyalprazolam, respectively. Plasma concentration ratios collected between 1 h and 48 h for both alprazolam/4-hydroxyalprazolam and alprazolam/alpha-hydroxyalprazolam correlated significantly to the corresponding AUC0-infinity ratios. The metabolic ratios of alprazolam to respective metabolite in a single plasma sample at 3-24 h are suggested to reflect the alprazolam 4- and alpha-hydroxylation activities. In future, it will be important to study these activities in populations where CYP3A5, in addition to CYP3A4, is expressed at a high frequency and to clarify the relative importance of the two enzymatic pathways for in vivo clearance of alprazolam.
Product Description
Supelco
alpha-Hydroxyalprazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®