MilliporeSigma
  • Antibacterial and antifungal activity of pinosylvin, a constituent of pine.
Antibacterial and antifungal activity of pinosylvin, a constituent of pine.
Fitoterapia (2005-03-09)
S K Lee, H J Lee, H Y Min, E J Park, K M Lee, Y H Ahn, Y J Cho, J H Pyee
ABSTRACT
The antibacterial and antifungal activities of pinosylvin (3,5-dihydroxy-trans-stilbene), a constituent of pine, were studied and compared with those of resveratrol (3,5,4'-trihydroxy-trans-stilbene). Pinosylvin exhibited more potent growth inhibitory activity against Candida albicans and Saccharomyces cerevisiae.
Sigma-Aldrich
Pinosylvin, ≥97.0% (HPLC)