MilliporeSigma
  • Protein heterodimerization through ligand-bridged multivalent pre-organization: enhancing ligand binding toward both protein targets.
Protein heterodimerization through ligand-bridged multivalent pre-organization: enhancing ligand binding toward both protein targets.
Journal of the American Chemical Society (2005-02-17)
Jiyun Liu, Zhongsheng Zhang, Xiaojian Tan, Wim G J Hol, Christophe L M J Verlinde, Erkang Fan
ABSTRACT
Structure-based design of a bifunctional ligand for two protein pentamers, cholera toxin B pentamer (CTB) and human serum amyloid P component (SAP), leads to multivalent dimerization of CTB and SAP in solution. This multivalent heterodimerization of proteins significantly enhances the affinity of the bifunctional ligand toward both target proteins.
Sigma-Aldrich
N-Acetyl-L-proline